CAS RN 68520-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Acetyl-6-(dimethylamino)naphthalene, TRC
CAS: 68520-00-3 Molecular Formula: C14 H15 N O Molecular Weight (g/mol): 213.28 IUPAC Name: 1-[6-(dimethylamino)naphthalen-2-yl]ethanone SMILES: CN(C)c1ccc2cc(ccc2c1)C(=O)C