accessibility menu, dialog, popup

UserName

CH3NNHSOOCH3(E/Z)

CAS RN 69015-74-3

IUPAC Name: 4-methyl-N'-(1-phenylpentylidene)benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-(1-phenylpentylidene)benzenesulfonohydrazide
Molecular Weight (g/mol): 330.45
Molecular Formula: C18H22N2O2S
InChi Key: VTEOESQGMPJHGP-UHFFFAOYSA-N
SMILES: CCCCC(=NNS(=O)(=O)C1=CC=C(C)C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results