accessibility menu, dialog, popup

UserName

CAS RN 690632-05-4

CH3CH3H3COONNHOOBr

CAS RN 690632-05-4

IUPAC Name: tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro[1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
Synonyms: tert-butyl 6-bromo-4-oxo-3H-spiro[1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
Molecular Weight (g/mol): 397.27
Molecular Formula: C17H21BrN2O4
InChi Key: FKBBJWUNJKLMBK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C1=C(O2)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results