accessibility menu, dialog, popup

UserName

CH3NCH3NOH

CAS RN 690632-68-9

IUPAC Name: [6-(diethylamino)pyridin-3-yl]methanol
Synonyms: [6-(diethylamino)pyridin-3-yl]methanol
Molecular Weight (g/mol): 180.25
Molecular Formula: C10H16N2O
InChi Key: OMGIPGLZKJOJSG-UHFFFAOYSA-N
SMILES: CCN(CC)C1=NC=C(CO)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results