accessibility menu, dialog, popup

UserName

CAS RN 6937-03-7

CH3OONH2N

CAS RN 6937-03-7

IUPAC Name: methyl 2-aminopyridine-4-carboxylate
Synonyms: methyl 2-aminopyridine-4-carboxylate
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: SVWWNEYBEFASMP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=NC(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results