accessibility menu, dialog, popup

UserName

CAS RN 694-05-3

HN

CAS RN 694-05-3

IUPAC Name: 1,2,3,6-tetrahydropyridine
Synonyms: 1,2,3,6-tetrahydropyridine delta(3)-piperideine 3-piperideine delta3-piperidine
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: FTAHXMZRJCZXDL-UHFFFAOYSA-N
SMILES: C1CC=CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results