accessibility menu, dialog, popup

UserName

CAS RN 694-80-4

ClBr

CAS RN 694-80-4

IUPAC Name: 1-bromo-2-chlorobenzene
Synonyms: 2-bromochlorobenzene
Molecular Weight (g/mol): 191.45
Molecular Formula: C6H4BrCl
InChi Key: QBELEDRHMPMKHP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)