accessibility menu, dialog, popup

UserName

CAS RN 6946-22-1

HClNH2OHOOHO

CAS RN 6946-22-1

IUPAC Name: hydrogen 3-aminobenzene-1,2-dicarboxylic acid chloride
Synonyms: hydrogen 3-aminobenzene-1,2-dicarboxylic acid chloride
Molecular Weight (g/mol): 217.61
Molecular Formula: C8H8ClNO4
InChi Key: ZBZAVEORKXFUQB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC(C(O)=O)=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results