accessibility menu, dialog, popup

UserName

HOO

CAS RN 6954-55-8

IUPAC Name: 9H-fluorene-4-carboxylic acid
Synonyms: 9H-fluorene-4-carboxylic acid
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: WJNBLUOXTBTGMB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1C1=CC=CC=C1C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)