accessibility menu, dialog, popup

UserName

CAS RN 696-04-8

CH3NHOHNO

CAS RN 696-04-8

IUPAC Name: 5-methyl-1,3-diazinane-2,4-dione
Synonyms: dihydrothymine 5-methyl-hydrouracil 5-methyl- (8CI) hydrouracil hydrouracil, 5-methyl- (8CI)
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: NBAKTGXDIBVZOO-UHFFFAOYNA-N
SMILES: CC1CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)