accessibility menu, dialog, popup

UserName

CAS RN 6966-09-2

ClON

CAS RN 6966-09-2

IUPAC Name: 2-[(piperidin-1-yl)methyl]cyclohexan-1-one hydrochloridyl
Synonyms: pimeclone hydrochloridyl
Molecular Weight (g/mol): 230.76
Molecular Formula: C12H21ClNO
InChi Key: MFMOSYZXKAREGX-UHFFFAOYNA-N
SMILES: [Cl].O=C1CCCCC1CN1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results