accessibility menu, dialog, popup

UserName

CH3NNH2NN

CAS RN 698-29-3

IUPAC Name: 4-amino-2-methylpyrimidine-5-carbonitrile
Synonyms: 4-amino-2-methylpyrimidine-5-carbonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: YBPNIILOUYAGIF-UHFFFAOYSA-N
SMILES: CC1=NC=C(C#N)C(N)=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)