accessibility menu, dialog, popup

UserName

CAS RN 69950-65-8

CH3OONO

CAS RN 69950-65-8

IUPAC Name: methyl 6-formylpyridine-2-carboxylate
Synonyms: methyl 6-formylpyridine-2-carboxylate
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: CERBENZCBVYKEF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results