accessibility menu, dialog, popup

UserName

CAS RN 70-00-8

IUPAC Name: 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: trifluridine
Molecular Weight (g/mol): 296.2
Molecular Formula: C10H11F3N2O5
InChi Key: VSQQQLOSPVPRAZ-UHFFFAOYNA-N
SMILES: OCC1OC(CC1O)N1C=C(C(=O)NC1=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results