CAS RN 7006-52-2
CAS RN 7006-52-2
IUPAC Name:
3-chloro-N-methylaniline
Synonyms:
3-chloro-N-methylaniline
Molecular Weight (g/mol):
141.6
Molecular Formula:
C7H8ClN
InChi Key:
WFGYSQDPURFIFL-UHFFFAOYSA-N
SMILES:
CNC1=CC=CC(Cl)=C1
1
–
2
of
2
results
2
3-Chloro-N-methylaniline, 97%
CAS: 7006-52-2 Molecular Formula: C7H8ClN Molecular Weight (g/mol): 141.60 MDL Number: MFCD00052014 InChI Key: WFGYSQDPURFIFL-UHFFFAOYSA-N Synonym: n-methyl-3-chloroaniline,3-chloro-n-methylbenzenamine,benzenamine, 3-chloro-n-methyl,3-chloro-phenyl-methyl-amine,n-methyl-m-chloroaniline,n1-methyl-3-chloroaniline,pubchem9202,acmc-209ocl,3-chloro-1-methylaniline,3-chloro-n-methyl aniline PubChem CID: 138900 IUPAC Name: 3-chloro-N-methylaniline SMILES: CNC1=CC=CC(Cl)=C1