CAS RN 7013-16-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Arainosine, TRC
CAS: 7013-16-3 Molecular Formula: C10H12N4O5 Molecular Weight (g/mol): 268.23 Synonym: 9-beta-D-Arabinofuranosyl-1,9-dihydro-6H-purin-6-one,9-beta-D-Arabinofuranosylhypoxanthine,Ara-H,Arabinosylhypoxanthine,Hypoxanthine Arabinoside,NSC 405122 IUPAC Name: 9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one SMILES: OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)n2cnc3C(=O)NC=Nc23