accessibility menu, dialog, popup

UserName

OHONOO

CAS RN 704-13-2

IUPAC Name: 3-hydroxy-4-nitrobenzaldehyde
Synonyms: 3-hydroxy-4-nitrobenzaldehyde
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: AUBBVPIQUDFRQI-UHFFFAOYSA-N
SMILES: OC1=CC(C=O)=CC=C1[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results