accessibility menu, dialog, popup

UserName

ONOClOCl

CAS RN 7073-36-1

IUPAC Name: 2-chloro-4-nitrobenzoyl chloride
Synonyms: 2-chloro-4-nitrobenzoyl chloride
Molecular Weight (g/mol): 220.01
Molecular Formula: C7H3Cl2NO3
InChi Key: KTHNITVDTYAHFF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results