CAS RN 7085-85-0
CAS RN 7085-85-0
IUPAC Name:
ethyl 2-cyanoprop-2-enoate
Synonyms:
ethyl cyanoacrylate
Molecular Weight (g/mol):
125.13
Molecular Formula:
C6H7NO2
InChi Key:
FGBJXOREULPLGL-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=C)C#N
1
–
2
of
2
results
2
Ethyl 2-Cyanoacrylate, TRC
CAS: 7085-85-0 Molecular Formula: C6 H7 N O2 Molecular Weight (g/mol): 125.13 Synonym: 2-Cyano-2-propenoic Acid Ethyl Ester,2-Cyanoacrylic Acid Ethyl Ester,910EM,ACE-E 50,ACE-EE,Adhesive 502,Anacure 3020,Loctite E 406,Sicomet 5195,Sicomet 8400,TB 1743,TK 200; IUPAC Name: ethyl 2-cyanoprop-2-enoate SMILES: CCOC(=O)C(=C)C#N