accessibility menu, dialog, popup

UserName

CAS RN 715-33-3

CH3BrHOBrOHOOCH3

CAS RN 715-33-3

IUPAC Name: methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
Synonyms: methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
Molecular Weight (g/mol): 339.97
Molecular Formula: C9H8Br2O4
InChi Key: OKALYLLBFOYLQB-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C(Br)=C(O)C(Br)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results