accessibility menu, dialog, popup

UserName

CAS RN 71830-07-4

NH3(R)(S)OO

CAS RN 71830-07-4

IUPAC Name: (1S,3R)-3-azaniumylcyclopentane-1-carboxylate
Synonyms: (1S,3R)-3-ammoniocyclopentane-1-carboxylate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: MLLSSTJTARJLHK-CRCLSJGQSA-N
SMILES: [NH3+][C@@H]1CC[C@@H](C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)