CAS RN 7232-21-5
CAS RN 7232-21-5
IUPAC Name:
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride
Synonyms:
maxolon
Molecular Weight (g/mol):
336.26
Molecular Formula:
C14H23Cl2N3O2
InChi Key:
RVFUNJWWXKCWNS-UHFFFAOYSA-N
SMILES:
Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
1
–
2
of
2
results
1
Metoclopramide hydrochloride, 98%
CAS: 7232-21-5 Molecular Formula: C14H23Cl2N3O2 Molecular Weight (g/mol): 336.26 InChI Key: RVFUNJWWXKCWNS-UHFFFAOYSA-N IUPAC Name: 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride SMILES: Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
2
Metoclopramide hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality