accessibility menu, dialog, popup

UserName

H3COOOOOOO

CAS RN 7241-98-7

IUPAC Name: 11-methoxy-6,8,16,20-tetraoxapentacyclo[10.8.0.0²,⁹.0³,⁷.0¹³,¹⁸]icosa-1,9,11,13(18)-tetraene-17,19-dione
Synonyms: aflatoxin G2
Molecular Weight (g/mol): 330.29
Molecular Formula: C17H14O7
InChi Key: WPCVRWVBBXIRMA-UHFFFAOYNA-N
SMILES: COC1=C2C3=C(C(=O)OCC3)C(=O)OC2=C2C3CCOC3OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results