CAS RN 72810-57-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
[[4-[(3-Methylphenyl)amino]-3-pyridinyl]sulfonyl]carbamic Acid Ethyl Ester, TRC
CAS: 72810-57-2 Molecular Formula: C15 H17 N3 O4 S Molecular Weight (g/mol): 335.38 Synonym: Ethyl [[4-[(3-Methylphenyl)amino]pyridin-3-yl]sulfonyl]carbamate,Torasemide, Anhydrous Imp. E (EP) IUPAC Name: ethyl N-[4-(3-methylanilino)pyridin-3-yl]sulfonylcarbamate SMILES: CCOC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2
2
Torasemide EP Impurity E, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.