accessibility menu, dialog, popup

UserName

CAS RN 72814-85-8

CH3CH3CH3CH3(E)(E)

CAS RN 72814-85-8

IUPAC Name: 4,4'-bis[(1E)-2-(2,5-dimethylphenyl)ethenyl]-1,1'-biphenyl
Synonyms: 4,4'-bis[(1E)-2-(2,5-dimethylphenyl)ethenyl]-1,1'-biphenyl
Molecular Weight (g/mol): 414.59
Molecular Formula: C32H30
InChi Key: AMDSCXLLKXHWOJ-IWGRKNQJSA-N
SMILES: CC1=CC=C(C)C(\C=C\C2=CC=C(C=C2)C2=CC=C(\C=C\C3=CC(C)=CC=C3C)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results