CAS RN 7283-09-2
CAS RN 7283-09-2
IUPAC Name:
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms:
d-galactose D-hexose D-hexoses glucose oxidase
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES:
OCC1OC(O)C(O)C(O)C1O
1
–
1
of
1
results
1
beta-D-Allopyranose, 97%, Thermo Scientific™
CAS: 7283-09-2 MDL Number: MFCD00063268