CAS RN 72867-72-2
CAS RN 72867-72-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2'-2'-Chloro-2-phenylacetophenone, TRC
CAS: 72867-72-2 Molecular Formula: C14H11ClO Molecular Weight (g/mol): 230.69 IUPAC Name: 1-(2-chlorophenyl)-2-phenylethanone SMILES: Clc1ccccc1C(=O)Cc2ccccc2