CAS RN 72918-21-9
CAS RN 72918-21-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,2,3,7,8,9-Hexachlorodibenzofuran, TRC
CAS: 72918-21-9 Molecular Formula: C12 H2 Cl6 O Molecular Weight (g/mol): 374.86 Synonym: Dibenzofuran, 1,2,3,7,8,9-hexachloro-,1,2,3,7,8,9-Hexachlorodibenzofuran,1,2,3,7,8,9-HxCDF,F 124,PCDF 124,1,2,3,7,8,9-HexaCDF,123789-HexaCDF,123789-HxCDF,123789-Hexachlorodibenzofuran IUPAC Name: 1,2,3,7,8,9-hexachlorodibenzofuran SMILES: Clc1cc2oc3cc(Cl)c(Cl)c(Cl)c3c2c(Cl)c1Cl