accessibility menu, dialog, popup

UserName

CAS RN 72956-09-3

CH3OONHOHONH

CAS RN 72956-09-3

IUPAC Name: 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
Synonyms: carvedilol carvedilol, (+-)-isomer (R)-carvedilol carvedilol, (S)-isomer (s)-carvedilol 14C-labeled carvedilol querto (r)-isomer carvedilol kredex carvedilol, 14c-labeled
Molecular Weight (g/mol): 406.48
Molecular Formula: C24H26N2O4
InChi Key: OGHNVEJMJSYVRP-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Grade 
  • (2)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)