CAS RN 7321-53-1
CAS RN 7321-53-1
IUPAC Name:
iron(3+) tris(2-ethylhexanoate)
Synonyms:
iron(3+) tris(2-ethylhexanoate)
Molecular Weight (g/mol):
485.46
Molecular Formula:
C24H45FeO6
InChi Key:
ULUYRVWYCIOFRV-UHFFFAOYNA-K
SMILES:
[Fe+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
1
–
1
of
1
results
1
Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%, Thermo Scientific™
CAS: 7321-53-1 Molecular Formula: C24H45FeO6 Molecular Weight (g/mol): 485.463 MDL Number: MFCD00014006 InChI Key: ULUYRVWYCIOFRV-UHFFFAOYSA-K Synonym: ferric 2-ethylhexanoate,iron 2-ethylhexanoate,iron tris 2-ethylhexanoate,iron ethylhexanoate,iron iii 2-ethylhexanoate,hexanoic acid, 2-ethyl-, iron 3+ salt 3:1,iron iii 2-ethylhexanoate nominally in mineral spirits, fe,iron tallate 2-ethylhexanoate,iron 3+ tris 2-ethylhexanoate,hexanoic acid, 2-ethyl-, iron 3+ salt PubChem CID: 110963 IUPAC Name: 2-ethylhexanoate;iron(3+) SMILES: CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3]