accessibility menu, dialog, popup

UserName

CAS RN 73220-38-9

ClOOBr

CAS RN 73220-38-9

IUPAC Name: 3-bromo-6-chloro-4H-chromen-4-one
Synonyms: 3-bromo-6-chlorochromen-4-one
Molecular Weight (g/mol): 259.48
Molecular Formula: C9H4BrClO2
InChi Key: ZILSBPAUJJBEFF-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC=C(Br)C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results