accessibility menu, dialog, popup

UserName

BrIN

CAS RN 73290-22-9

IUPAC Name: 2-bromo-5-iodopyridine
Synonyms: 2-bromo-5-iodopyridine
Molecular Weight (g/mol): 283.89
Molecular Formula: C5H3BrIN
InChi Key: LLKRSJVPTKFSLS-UHFFFAOYSA-N
SMILES: BrC1=CC=C(I)C=N1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)