CAS RN 73364-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(n-butylcyclopentadienyl)zirconium(IV) dichloride, 98+%, Thermo Scientific Chemicals
CAS: 73364-10-0 Molecular Formula: C18H26Cl2Zr MDL Number: MFCD00672105 Synonym: 1,1'-Di-n-butylzirconocene dichloride