accessibility menu, dialog, popup

UserName

CH3NH3COBr(E/Z)

CAS RN 73387-60-7

IUPAC Name: 1-(4-bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Synonyms: 1-(4-bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Molecular Weight (g/mol): 254.13
Molecular Formula: C11H12BrNO
InChi Key: IGZLESKZUATMSD-UHFFFAOYSA-N
SMILES: CN(C)C=CC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results