accessibility menu, dialog, popup

UserName

CAS RN 73576-33-7

H3CCH3NNH2NCl

CAS RN 73576-33-7

IUPAC Name: 4-chloro-6-(propan-2-yl)pyrimidin-2-amine
Synonyms: 4-chloro-6-isopropylpyrimidin-2-amine
Molecular Weight (g/mol): 171.63
Molecular Formula: C7H10ClN3
InChi Key: SPMMDFHRMVXLLO-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(Cl)=NC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results