accessibility menu, dialog, popup

UserName

CH3ONHONH2(E/Z)

CAS RN 73647-50-4

IUPAC Name: N'-hydroxy-3-methoxybenzene-1-carboximidamide
Synonyms: N'-hydroxy-3-methoxybenzenecarboximidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: CEHMGZTZNNEADY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results