accessibility menu, dialog, popup

UserName

CAS RN 73963-72-1

OONNNNHN

CAS RN 73963-72-1

IUPAC Name: 6-[4-(1-cyclohexyl-1H-1,2,3,4-tetrazol-5-yl)butoxy]-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: cilostazol cilostazole pletal cilostazol (substance) otsuka brand of cilostazol pharmacia brand of cilostazol cilostazol (product) pletaal cilostazolum cilostazol preparation
Molecular Weight (g/mol): 369.47
Molecular Formula: C20H27N5O2
InChi Key: RRGUKTPIGVIEKM-UHFFFAOYSA-N
SMILES: O=C1CCC2=CC(OCCCCC3=NN=NN3C3CCCCC3)=CC=C2N1
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)