CAS RN 74115-10-9
CAS RN 74115-10-9
IUPAC Name:
6-chloro-1-(3-methylphenyl)-3-(prop-2-en-1-yl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Synonyms:
skf 83822 hydrobromide
Molecular Weight (g/mol):
424.76
Molecular Formula:
C20H23BrClNO2
InChi Key:
CFWPKYBBXBANLU-UHFFFAOYNA-N
SMILES:
Br.CC1=CC=CC(=C1)C1CN(CC=C)CCC2=C(Cl)C(O)=C(O)C=C12
1
–
1
of
1
results
1
Tocris Bioscience™ SKF 83822 hydrobromide
CAS: 74115-10-9 Molecular Formula: C20H23BrClNO2