accessibility menu, dialog, popup

UserName

H3CSNOO

CAS RN 74470-23-8

IUPAC Name: 2-(methylsulfanyl)pyridine-3-carboxylate
Synonyms: 2-(methylsulfanyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 168.19
Molecular Formula: C7H6NO2S
InChi Key: COPSJQVPEUUOKY-UHFFFAOYSA-M
SMILES: CSC1=NC=CC=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results