CAS RN 74578-69-1
CAS RN 74578-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ceftriaxone sodium salt, MedChemExpress
MedChemExpress Ceftriaxone sodium salt (Disodium ceftriaxone) is a third-generation cephalosporin antibiotic with excellent activity against many gram-negative, and reasonable activity against most gram-positive microorganisms. Anti-inflammatory and antioxidant characteristics.