accessibility menu, dialog, popup

UserName

CAS RN 745784-04-7

NFOHOF

CAS RN 745784-04-7

IUPAC Name: 3,5-difluoropyridine-2-carboxylic acid
Synonyms: 3,5-difluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: QKLXAJQKMIWFRC-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(F)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results