CAS RN 74602-32-7
IUPAC Name:
dodeca-5,7-diyne-1,12-diol
Synonyms:
dodeca-5,7-diyne-1,12-diol
Molecular Weight (g/mol):
194.27
Molecular Formula:
C12H18O2
InChi Key:
SEWYHOMCDKWYEF-UHFFFAOYSA-N
SMILES:
OCCCCC#CC#CCCCCO
1
–
1
of
1
results
1
5,7-Dodecadiyne-1,12-diol, 96%, Thermo Scientific™
CAS: 74602-32-7 Molecular Formula: C12H18O2 Molecular Weight (g/mol): 194.27 MDL Number: MFCD00078343 InChI Key: SEWYHOMCDKWYEF-UHFFFAOYSA-N Synonym: 5,7-dodecadiyne-1,12-diol,5,7-dodecadiyn-1,12-diol,5,7-dodecadiyne-1,12-diol #,5,7-dodecadiyn-1,12-diol, PubChem CID: 560878 IUPAC Name: dodeca-5,7-diyne-1,12-diol SMILES: OCCCCC#CC#CCCCCO