CAS RN 747413-08-7
CAS RN 747413-08-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
VER-50589, MedChemExpress
MedChemExpress VER-50589 is a Hsp90 inhibitor, with an IC50 of 21 nM and a Kd of 4.5 nM.