accessibility menu, dialog, popup

UserName

CH3NH3COF(E/Z)

CAS RN 75175-77-8

IUPAC Name: 3-(dimethylamino)-1-(4-fluorophenyl)prop-2-en-1-one
Synonyms: 3-(dimethylamino)-1-(4-fluorophenyl)prop-2-en-1-one
Molecular Weight (g/mol): 193.22
Molecular Formula: C11H12FNO
InChi Key: GTJNUCRUOWBWEW-UHFFFAOYSA-N
SMILES: CN(C)C=CC(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results