CAS RN 752222-83-6
CAS RN 752222-83-6
IUPAC Name:
3-ethyl-1-propyl-8-(1-{[3-(trifluoromethyl)phenyl]methyl}-1H-pyrazol-4-yl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms:
3-ethyl-1-propyl-8-(1-{[3-(trifluoromethyl)phenyl]methyl}pyrazol-4-yl)-7H-purine-2,6-dione
Molecular Weight (g/mol):
446.43
Molecular Formula:
C21H21F3N6O2
InChi Key:
KOYXXLLNCXWUNF-UHFFFAOYSA-N
SMILES:
CCCN1C(=O)N(CC)C2=C(NC(=N2)C2=CN(CC3=CC(=CC=C3)C(F)(F)F)N=C2)C1=O