accessibility menu, dialog, popup

UserName

HOFF

CAS RN 75853-20-2

IUPAC Name: (2,5-difluorophenyl)methanol
Synonyms: (2,5-difluorophenyl)methanol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: KIKCAPPVFQLOIU-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results