accessibility menu, dialog, popup

UserName

CAS RN 76116-20-6

H3C(R)OHBr

CAS RN 76116-20-6

IUPAC Name: (1R)-1-(2-bromophenyl)ethan-1-ol
Synonyms: (1R)-1-(2-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: DZLZSFZSPIUINR-ZCFIWIBFSA-N
SMILES: C[C@@H](O)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results