accessibility menu, dialog, popup

UserName

CAS RN 76347-13-2

CH3H3CBOH3CH3CH3CCH3O

CAS RN 76347-13-2

IUPAC Name: 4,4,5,5-tetramethyl-2-(propan-2-yl)-1,3,2-dioxaborolane
Synonyms: 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 170.06
Molecular Formula: C9H19BO2
InChi Key: MECCSFDHAVAAAW-UHFFFAOYSA-N
SMILES: CC(C)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results