accessibility menu, dialog, popup

UserName

CAS RN 76530-12-6

IUPAC Name: naphthalene-2-sulfonic acid hydrate
Synonyms: 2-naphthalenesulfonic acid hydrate
Molecular Weight (g/mol): 226.25
Molecular Formula: C10H10O4S
InChi Key: HKGOFWIZZIYCOS-UHFFFAOYSA-N
SMILES: O.OS(=O)(=O)C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)