accessibility menu, dialog, popup

UserName

CAS RN 76579-64-1

IUPAC Name: [3-(4-bromophenoxy)propyl]dimethylamine
Synonyms: [3-(4-bromophenoxy)propyl]dimethylamine
Molecular Weight (g/mol): 258.16
Molecular Formula: C11H16BrNO
InChi Key: CAPNMXRLKVMOOB-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results